Assay Details
Gene Symbol: Srf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109302.1 | NP_001102772.1 | 3 - 4 | 1364 | 78 | |
XM_006244557.2 | XP_006244619.1 | 3 - 4 | 1367 | 78 | ||
XM_006244558.2 | XP_006244620.1 | 3 - 4 | 1368 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
