Assay Details
Gene Symbol: LOC100911260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006229638.1 | XP_006229700.1 | 3 - 4 | 345 | 80 |
Gene Symbol: Hcfc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001139507.1 | NP_001132979.1 | 14 - 15 | 3614 | 80 | |
XM_006229593.2 | XP_006229655.1 | 14 - 15 | 3593 | 80 | ||
XM_006229594.2 | XP_006229656.1 | 14 - 15 | 3592 | 80 | ||
XM_006229596.2 | XP_006229658.1 | 14 - 15 | 3590 | 80 | ||
XM_006229597.1 | XP_006229659.1 | 14 - 15 | 3609 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
