Assay Details
Gene Symbol: Tpmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006253770.2 | XP_006253832.1 | 5 - 6 | 568 | 68 | |
GenBank mRNA | AF120100.1 | - | 4 - 5 | 408 | 68 | |
BC158556.1 | - | 5 - 6 | 474 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
