Assay Details
Gene Symbol: Tpmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079531.1 | NP_001072999.1 | 6 - 7 | 573 | 74 | |
XM_006253770.2 | XP_006253832.1 | 6 - 7 | 636 | 74 | ||
GenBank mRNA | AF120100.1 | - | 5 - 6 | 476 | 74 | |
AY325236.1 | - | 6 - 7 | 573 | 74 | ||
BC158556.1 | - | 6 - 7 | 542 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
