Assay Details
Gene Symbol: Tpmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079531.1 | NP_001072999.1 | 8 - 9 | 708 | 75 | |
XM_006253770.2 | XP_006253832.1 | 8 - 9 | 771 | 75 | ||
GenBank mRNA | AF120100.1 | - | 7 - 8 | 611 | 75 | |
AY325236.1 | - | 8 - 9 | 708 | 75 | ||
BC158556.1 | - | 8 - 9 | 677 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
