Assay Details
Gene Symbol: Cdyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014145.1 | NP_001014167.1 | 7 - 8 | 1519 | 136 | |
XM_006253839.2 | XP_006253901.1 | 5 - 6 | 1709 | 136 | ||
XM_006253840.2 | XP_006253902.1 | 5 - 6 | 1569 | 136 | ||
GenBank mRNA | BC079003.1 | - | 7 - 8 | 1519 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
