Assay Details
Gene Symbol: Ror2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107339.1 | NP_001100809.1 | 6 - 7 | 1142 | 86 | |
XM_008771495.1 | XP_008769717.1 | 5 - 6 | 854 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
