Assay Details
Gene Symbol: Ror2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107339.1 | NP_001100809.1 | 7 - 8 | 1380 | 71 | |
XM_008771495.1 | XP_008769717.1 | 6 - 7 | 1092 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
