Assay Details
Gene Symbol: Mocs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007633.2 | NP_001007634.1 | 1 - 2 | 94 | 70 | |
XM_006231986.2 | XP_006232048.1 | 1 - 2 | 92 | 70 | ||
GenBank mRNA | FM123325.1 | - | 1 - 2 | 48 | 70 | |
FQ220210.1 | - | 1 - 2 | 40 | 70 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001007633.2, XM_006231986.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
