Assay Details
Gene Symbol: Park2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_020093.1 | NP_064478.1 | 4 - 5 | 608 | 73 | |
GenBank mRNA | AB039878.1 | - | 4 - 5 | 608 | 73 | |
AF168004.1 | - | 4 - 5 | 628 | 73 | ||
AF210434.1 | - | 4 - 5 | 600 | 73 | ||
AF257234.1 | - | 3 - 4 | 538 | 73 | ||
AF343574.1 | - | 4 - 5 | 297 | 73 | ||
AF343575.1 | - | 2 - 3 | 374 | 73 | ||
AF381277.1 | - | 4 - 5 | 610 | 73 | ||
AF381278.1 | - | 3 - 4 | 538 | 73 | ||
AF381280.1 | - | 4 - 5 | 694 | 73 | ||
GU322017.1 | - | 5 - 6 | 707 | 73 | ||
GU322364.1 | - | 4 - 5 | 551 | 73 | ||
GU345836.1 | - | 4 - 5 | 551 | 73 | ||
KC774169.1 | - | 3 - 4 | 394 | 73 | ||
KC774170.1 | - | 3 - 4 | 394 | 73 | ||
KC774172.1 | - | 4 - 5 | 548 | 73 | ||
KC774173.1 | - | 3 - 4 | 394 | 73 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AF343574.1,AF343575.1,AF381280.1,KC774169.1,KC774170.1,KC774173.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
