Assay Details
Gene Symbol: Nipa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107518.1 | NP_001100988.1 | 4 - 5 | 680 | 88 | |
XM_006229292.2 | XP_006229354.1 | 6 - 7 | 783 | 88 | ||
XM_006229295.2 | XP_006229357.1 | 4 - 5 | 494 | 88 | ||
XM_006229296.2 | XP_006229358.1 | 4 - 5 | 550 | 88 | ||
XM_006229297.2 | XP_006229359.1 | 3 - 4 | 539 | 88 | ||
XM_006229298.2 | XP_006229360.1 | 3 - 4 | 357 | 88 | ||
XM_006229299.2 | XP_006229361.1 | 3 - 4 | 429 | 88 | ||
XM_008759390.1 | XP_008757612.1 | 3 - 4 | 385 | 88 | ||
XM_008759391.1 | XP_008757613.1 | 3 - 4 | 361 | 88 | ||
XM_008759392.1 | XP_008757614.1 | 5 - 6 | 944 | 88 | ||
GenBank mRNA | FQ222971.1 | - | 3 - 4 | 543 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
