Assay Details
Gene Symbol: Nipa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107518.1 | NP_001100988.1 | 5 - 6 | 773 | 148 | |
XM_006229292.2 | XP_006229354.1 | 7 - 8 | 876 | 148 | ||
XM_006229295.2 | XP_006229357.1 | 5 - 6 | 587 | 148 | ||
XM_006229296.2 | XP_006229358.1 | 5 - 6 | 643 | 148 | ||
XM_006229297.2 | XP_006229359.1 | 4 - 5 | 632 | 148 | ||
XM_006229298.2 | XP_006229360.1 | 4 - 5 | 450 | 148 | ||
XM_006229299.2 | XP_006229361.1 | 4 - 5 | 522 | 148 | ||
XM_008759390.1 | XP_008757612.1 | 4 - 5 | 478 | 148 | ||
XM_008759391.1 | XP_008757613.1 | 4 - 5 | 454 | 148 | ||
XM_008759392.1 | XP_008757614.1 | 6 - 7 | 1037 | 148 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
