Assay Details
Gene Symbol: Aldh5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022851.1 | NP_074042.1 | 1 - 2 | 339 | 99 | |
GenBank mRNA | FM032642.1 | - | 1 - 2 | 339 | 99 | |
L34821.1 | - | 1 - 2 | 218 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
