Assay Details
Gene Symbol: Traf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107815.2 | NP_001101285.1 | 2 - 3 | 310 | 80 | |
XM_006233594.2 | XP_006233656.1 | 3 - 4 | 370 | 80 | ||
XM_006233596.2 | XP_006233658.1 | 3 - 4 | 376 | 80 | ||
XM_008761588.1 | XP_008759810.1 | 3 - 4 | 316 | 80 | ||
GenBank mRNA | BC169064.1 | - | 2 - 3 | 310 | 5599352 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
