Assay Details
Gene Symbol: Gfi1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107823.1 | NP_001101293.1 | 3 - 4 | 360 | 79 | |
XM_006233789.1 | XP_006233851.1 | 3 - 4 | 399 | 79 | ||
XM_008761646.1 | XP_008759868.1 | 3 - 4 | 447 | 79 | ||
XM_008761647.1 | XP_008759869.1 | 3 - 4 | 3721 | 79 | ||
XM_008761648.1 | XP_008759870.1 | 4 - 5 | 611 | 79 | ||
XM_008761649.1 | XP_008759871.1 | 3 - 4 | 521 | 79 | ||
XM_008761650.1 | XP_008759872.1 | 3 - 4 | 999 | 79 | ||
XM_008761651.1 | XP_008759873.1 | 3 - 4 | 447 | 79 | ||
GenBank mRNA | BC158685.1 | - | 3 - 4 | 375 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
