Assay Details
Gene Symbol: Ttll11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001071863.5 | XP_001071863.3 | 4 - 5 | 1891 | 99 | |
GenBank mRNA | BC169049.1 | - | 2 - 3 | 721 | 7374372 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
