Assay Details
Gene Symbol: Ttll11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001071863.5 | XP_001071863.3 | 8 - 9 | 2449 | 111 | |
GenBank mRNA | BC169049.1 | - | 6 - 7 | 1279 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P148265 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P017495 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 331294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899045 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 749506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8006 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723965 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 243971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 348447 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168197 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642014 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 704480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 724466 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914618 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858236 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818930 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69636 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 45563 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P148265 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P017495 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P017495 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P148265 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P148265 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P017495 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P379987 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10844725 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10844710 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10844711 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10844712 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10844716 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10844709 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10844721 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10844714 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10844718 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10844724 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10844715 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10844708 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10844707 | - | - |
Rat Genome 230 2.0 Array | 1380266_PM_at | - | - |
Rat Genome 230 2.0 Array | 1393748_at | - | - |
Rat Genome 230 2.0 Array | 1380266_at | - | - |
Rat Genome 230 2.0 Array | 1393748_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1380266_at | - | - |
Affymetrix HT Rat Focus Array | 1380266_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300022955.rn.1 | TC0300002942.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300022956.rn.1 | TC0300002942.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300022954.rn.1 | TC0300002942.rn.1 | RTA TC |
Rat Expression Set 230 | 1393748_at | - | - |
Rat Expression Set 230 | 1380266_at | - | - |
Rat Expression Set 230 | 1393748_PM_at | - | - |
Rat Expression Set 230 | 1380266_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10844719 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10844717 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10844707 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21114412 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1356438 | - | - |
