Assay Details
Gene Symbol: Crat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004085.2 | NP_001004085.1 | 9 - 10 | 1425 | 141 | |
XM_006233823.1 | XP_006233885.1 | 10 - 11 | 1945 | 141 | ||
XM_008761663.1 | XP_008759885.1 | 10 - 11 | 1428 | 141 | ||
XM_008761664.1 | XP_008759886.1 | 9 - 10 | 1294 | 141 | ||
GenBank mRNA | AJ620886.1 | - | 9 - 10 | 1203 | 141 | |
BC083616.1 | - | 9 - 10 | 1425 | 7133847 | 141 | |
FQ230824.1 | - | 10 - 11 | 1540 | 141 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
