Assay Details
Gene Symbol: Exosc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108952.1 | NP_001102422.1 | 4 - 5 | 378 | 58 | |
GenBank mRNA | FQ227372.1 | - | 4 - 5 | 370 | 58 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
