Assay Details
Gene Symbol: Anapc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100532.1 | NP_001094002.1 | 10 - 11 | 1932 | 69 | |
GenBank mRNA | BC107471.1 | - | 8 - 9 | 1036 | 69 | |
BC166796.1 | - | 10 - 11 | 1932 | 7938000 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
