Assay Details
Gene Symbol: Anapc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100532.1 | NP_001094002.1 | 5 - 6 | 1216 | 86 | |
GenBank mRNA | BC107471.1 | - | 3 - 4 | 320 | 86 | |
BC166796.1 | - | 5 - 6 | 1216 | 7938000 | 86 | |
FQ224461.1 | - | 5 - 6 | 1226 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
