Assay Details
Gene Symbol: Anapc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100532.1 | NP_001094002.1 | 7 - 8 | 1513 | 94 | |
GenBank mRNA | BC107471.1 | - | 5 - 6 | 617 | 94 | |
BC166796.1 | - | 7 - 8 | 1513 | 7938000 | 94 | |
FQ224461.1 | - | 7 - 8 | 1523 | 94 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
