Assay Details
Gene Symbol: Nacc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100533.1 | NP_001094003.1 | 4 - 5 | 1277 | 87 | |
XM_006233679.2 | XP_006233741.1 | 4 - 5 | 8376 | 87 | ||
XM_008761623.1 | XP_008759845.1 | 3 - 4 | 2480 | 87 | ||
GenBank mRNA | BC092608.1 | - | 3 - 4 | 1081 | 87 | |
FQ210633.1 | - | 4 - 5 | 686 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
