Assay Details
Gene Symbol: Nacc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100533.1 | NP_001094003.1 | 5 - 6 | 1378 | 101 | |
XM_006233679.2 | XP_006233741.1 | 5 - 6 | 8477 | 101 | ||
XM_008761623.1 | XP_008759845.1 | 4 - 5 | 2581 | 101 | ||
GenBank mRNA | BC092608.1 | - | 4 - 5 | 1182 | 101 | |
FQ210633.1 | - | 5 - 6 | 787 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
negative regulation of cell proliferation
posttranscriptional regulation of gene expression
histone deacetylation
cellular protein complex localization
negative regulation of transcription, DNA-templated
protein homooligomerization
negative regulation of G1/S transition of mitotic cell cycle by negative regulation of transcription from RNA polymerase II promoter
positive regulation of intrinsic apoptotic signaling pathway in response to DNA damage
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, RNA polymerase II transcription factor binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
histone deacetylase activity
protein homodimerization activity
histone deacetylase binding
transcription from RNA polymerase II promoter
negative regulation of cell proliferation
posttranscriptional regulation of gene expression
histone deacetylation
cellular protein complex localization
negative regulation of transcription, DNA-templated
protein homooligomerization
negative regulation of G1/S transition of mitotic cell cycle by negative regulation of transcription from RNA polymerase II promoter
positive regulation of intrinsic apoptotic signaling pathway in response to DNA damage
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, RNA polymerase II transcription factor binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
histone deacetylase activity
protein homodimerization activity
histone deacetylase binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P069646 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 907648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 655926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734101 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255107 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450993 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 578078 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 218345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 430550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393495 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501138 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 579584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 858287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496711 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797183 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 456404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 157615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 193652 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 373243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418649 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386023 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 428862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 837881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 156511 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 65277 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38886 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P069646 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P069646 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P069646 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P361144 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 30149 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10843653 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10843654 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10843652 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10843648 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10843649 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10843650 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10843647 | - | - |
Rat Genome 230 2.0 Array | 1380459_at | - | - |
Rat Genome 230 2.0 Array | 1380459_PM_at | - | - |
Rat Genome 230 2.0 Array | 1397967_PM_at | - | - |
Rat Genome 230 2.0 Array | 1397967_at | - | - |
Affymetrix HT Rat Focus Array | 1380459_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1380459_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0300020348.rn.1 | TC0300002710.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300020350.rn.1 | TC0300002710.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0300020349.rn.1 | TC0300002710.rn.1 | RTA TC |
Rat Expression Set 230 | 1380459_PM_at | - | - |
Rat Expression Set 230 | 1397967_PM_at | - | - |
Rat Expression Set 230 | 1380459_at | - | - |
Rat Expression Set 230 | 1397967_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10843647 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20806293 | - | - |
ABI Rat Genome Survey Microarray | 22313585 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371232 | - | - |
