Assay Details
Gene Symbol: Tada1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037980.2 | NP_001033069.2 | 2 - 3 | 264 | 69 | |
GenBank mRNA | BC091380.1 | - | 154 | 69 | ||
BC158713.1 | - | 2 - 3 | 264 | 69 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC091380.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
