Assay Details
Gene Symbol: Tada1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037980.2 | NP_001033069.2 | 7 - 8 | 957 | 113 | |
GenBank mRNA | BC091380.1 | - | 749 | 113 | ||
BC158713.1 | - | 7 - 8 | 957 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
