Assay Details
Gene Symbol: Rgs13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769582.1 | XP_008767804.1 | 3 - 4 | 451 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
