Assay Details
Gene Symbol: Nuak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007617.1 | NP_001007618.1 | 3 - 4 | 590 | 53 | |
XM_008769444.1 | XP_008767666.1 | 3 - 4 | 1291 | 53 | ||
GenBank mRNA | BC081899.1 | - | 3 - 4 | 590 | 53 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
