Assay Details
Gene Symbol: Axdnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769725.1 | XP_008767947.1 | 10 - 11 | 1561 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Axdnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008769725.1 | XP_008767947.1 | 10 - 11 | 1561 | 83 |
Set Membership: |
![]() ![]() ![]() |