Assay Details
Gene Symbol: Nr5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006249928.2 | XP_006249990.1 | 1 - 2 | 328 | 117 | |
GenBank mRNA | AB012961.1 | - | 1 - 2 | 157 | 117 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AB012961.1, XM_006249928.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
