Assay Details
Gene Symbol: Nr5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021742.1 | NP_068510.1 | 5 - 6 | 616 | 108 | |
XM_006249928.2 | XP_006249990.1 | 4 - 5 | 724 | 108 | ||
XM_006249929.2 | XP_006249991.1 | 3 - 4 | 464 | 108 | ||
GenBank mRNA | AB012960.1 | - | 5 - 6 | 616 | 108 | |
AB012961.1 | - | 4 - 5 | 553 | 108 | ||
U47280.1 | - | 2 - 3 | 270 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
