Assay Details
Gene Symbol: Tor1aip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165896.1 | NP_001159368.1 | 2 - 3 | 158 | 54 | |
GenBank mRNA | CO565924.1 | - | 196 | 54 | ||
CV107022.1 | - | 2 - 3 | 117 | 54 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001165896.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
