Assay Details
Gene Symbol: Arhgef37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100974.1 | NP_001094444.1 | 5 - 6 | 785 | 79 | |
XM_006254807.2 | XP_006254869.1 | 5 - 6 | 844 | 79 | ||
XM_006254808.2 | XP_006254870.1 | 5 - 6 | 2784 | 79 | ||
XM_006254809.2 | XP_006254871.1 | 5 - 6 | 751 | 79 | ||
XM_006254811.2 | XP_006254873.1 | 5 - 6 | 880 | 79 | ||
GenBank mRNA | AB199794.1 | - | 5 - 6 | 785 | 79 | |
BC168236.1 | - | 827 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
