Assay Details
Gene Symbol: Arhgef37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100974.1 | NP_001094444.1 | 12 - 13 | 1946 | 63 | |
XM_006254807.2 | XP_006254869.1 | 12 - 13 | 2005 | 63 | ||
XM_006254808.2 | XP_006254870.1 | 12 - 13 | 3945 | 63 | ||
XM_006254809.2 | XP_006254871.1 | 12 - 13 | 1912 | 63 | ||
XM_006254811.2 | XP_006254873.1 | 12 - 13 | 2041 | 63 | ||
GenBank mRNA | AB199794.1 | - | 12 - 13 | 1946 | 63 | |
BC168236.1 | - | 1988 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P066483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 939580 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 346661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117450 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 407393 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 860048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261985 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739379 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 952581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 338799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 541314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247557 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 5957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 217126 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232467 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772255 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 575939 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561008 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 608760 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 477994 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 848239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 684996 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 624448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745622 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 118804 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74236 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71834 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87900 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120779 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106515 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P066483 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P066483 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P066483 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P154445 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 96991 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10804695 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804704 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804703 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804702 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804701 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804699 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804707 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804696 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804706 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804698 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804700 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804709 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804697 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804694 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10804708 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10804693 | - | - |
Affymetrix Rat Clariom S Assay | PSR1800010762.rn.1 | TC1800001521.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1800010764.rn.1 | TC1800001521.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1800010763.rn.1 | TC1800001521.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1800010767.rn.1 | TC1800001521.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10804693 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21545635 | - | - |
ABI Rat Genome Survey Microarray | 21648683 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1356033 | - | - |
