Assay Details
Gene Symbol: Trim36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106147.1 | NP_001099617.1 | 3 - 4 | 657 | 110 | |
XM_006254686.2 | XP_006254748.1 | 3 - 4 | 885 | 110 | ||
XM_008772127.1 | XP_008770349.1 | 3 - 4 | 885 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
