Assay Details
Gene Symbol: Trim36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106147.1 | NP_001099617.1 | 4 - 5 | 806 | 84 | |
XM_006254686.2 | XP_006254748.1 | 4 - 5 | 1034 | 84 | ||
XM_008772127.1 | XP_008770349.1 | 4 - 5 | 1034 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
