Assay Details
Gene Symbol: Fech | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108434.1 | NP_001101904.1 | 6 - 7 | 797 | 98 | |
XM_006254850.2 | XP_006254912.1 | 6 - 7 | 3051 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
