Assay Details
Gene Symbol: Fech | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108434.1 | NP_001101904.1 | 9 - 10 | 1174 | 127 | |
XM_006254850.2 | XP_006254912.1 | 9 - 10 | 3428 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
