Assay Details
Gene Symbol: Malt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751798.3 | XP_003751846.1 | 14 - 15 | 1990 | 76 | |
XM_006254888.2 | XP_006254950.1 | 14 - 15 | 1990 | 76 | ||
XM_225927.8 | XP_225927.5 | 13 - 14 | 1956 | 76 | ||
GenBank mRNA | CO556697.1 | - | 1 - 2 | 159 | 76 | |
FQ225105.1 | - | 4 - 5 | 414 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
