Assay Details
Gene Symbol: Malt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751798.3 | XP_003751846.1 | 16 - 17 | 2278 | 110 | |
XM_225927.8 | XP_225927.5 | 15 - 16 | 2244 | 110 | ||
GenBank mRNA | CO556697.1 | - | 3 - 4 | 447 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
