Assay Details
Gene Symbol: Malt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751798.3 | XP_003751846.1 | 3 - 4 | 733 | 109 | |
XM_006254888.2 | XP_006254950.1 | 3 - 4 | 733 | 109 | ||
XM_225927.8 | XP_225927.5 | 3 - 4 | 732 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
