Assay Details
Gene Symbol: Bub3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047906.2 | NP_001041371.2 | 1 - 2 | 67 | 75 | |
GenBank mRNA | BC099199.1 | - | 1 - 2 | 41 | 75 | |
DY575212.1 | - | 1 - 2 | 67 | 75 | ||
FQ225106.1 | - | 1 - 2 | 47 | 75 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC099199.1, NM_001047906.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
