Assay Details
Gene Symbol: Bub3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047906.2 | NP_001041371.2 | 3 - 4 | 331 | 110 | |
GenBank mRNA | BC099199.1 | - | 3 - 4 | 305 | 110 | |
DY575212.1 | - | 3 - 4 | 331 | 110 | ||
FQ225106.1 | - | 3 - 4 | 311 | 110 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
