Assay Details
Gene Symbol: Rcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006231208.2 | XP_006231270.1 | 2 - 3 | 320 | 93 | |
GenBank mRNA | AY310155.1 | - | 397 | 93 | ||
AY325146.1 | - | 397 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
