Assay Details
Gene Symbol: Rcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013152.1 | NP_001013170.1 | 1 - 2 | 250 | 66 | |
XM_006231208.2 | XP_006231270.1 | 5 - 6 | 696 | 66 | ||
GenBank mRNA | BC091413.1 | - | 1 - 2 | 250 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
