Assay Details
Gene Symbol: Rcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013152.1 | NP_001013170.1 | 3 - 4 | 531 | 87 | |
XM_006231208.2 | XP_006231270.1 | 7 - 8 | 977 | 87 | ||
GenBank mRNA | AY310155.1 | - | 967 | 87 | ||
AY325146.1 | - | 967 | 87 | |||
BC091413.1 | - | 3 - 4 | 531 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
