Assay Details
Gene Symbol: Suv420h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108512.1 | NP_001101982.1 | 11 - 12 | 1870 | 85 | |
XM_008760150.1 | XP_008758372.1 | 8 - 9 | 1333 | 85 | ||
XM_008760151.1 | XP_008758373.1 | 9 - 10 | 1635 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
muscle organ development
biological_process
histone methylation
histone H4-K20 methylation
histone H4-K20 trimethylation
molecular_function
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H4-K20 specific)
transcription, DNA-templated
regulation of transcription, DNA-templated
muscle organ development
biological_process
histone methylation
histone H4-K20 methylation
histone H4-K20 trimethylation
molecular_function
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H4-K20 specific)
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P556491 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P556707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 606509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 842026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 623688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 591803 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 438739 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816428 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 393666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 64292 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 371125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205987 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 796977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725514 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454425 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734799 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 942386 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 372391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31819 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 129699 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P556491 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P556707 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P556491 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P556707 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P556707 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P556491 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P789584 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 6847 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10712696 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712695 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712698 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712701 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712697 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712705 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712702 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712703 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712700 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712704 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712699 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10712694 | - | - |
Rat Genome 230 2.0 Array | 1390198_at | - | - |
Rat Genome 230 2.0 Array | 1390198_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0100023962.rn.1 | TC0100002982.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0100023961.rn.1 | TC0100002982.rn.1 | RTA TC |
Rat Expression Set 230 | 1390198_at | - | - |
Rat Expression Set 230 | 1390198_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10712694 | - | - |
Rat Genome U34 Set | rc_AI012516_at | - | - |
Rat Genome U34 Set | rc_AI176517_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21452744 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1365857 | - | - |
