Assay Details
Gene Symbol: Fads1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053445.2 | NP_445897.2 | 9 - 10 | 1163 | 79 | |
XM_006231075.2 | XP_006231137.1 | 9 - 10 | 1310 | 79 | ||
GenBank mRNA | AB052085.1 | - | 9 - 10 | 1163 | 79 | |
AF320509.1 | - | 9 - 10 | 1192 | 79 | ||
FQ218734.1 | - | 8 - 9 | 1096 | 79 | ||
HQ909026.1 | - | 9 - 10 | 1087 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
