Assay Details
Gene Symbol: Il33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014166.1 | NP_001014188.1 | 7 - 8 | 671 | 64 | |
XM_008760344.1 | XP_008758566.1 | 7 - 8 | 709 | 64 | ||
GenBank mRNA | BC081993.1 | - | 7 - 8 | 671 | 7130307 | 64 |
FQ212643.1 | - | 7 - 8 | 699 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
