Assay Details
Gene Symbol: Slc3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271089.2 | NP_001258018.1 | 2 - 3 | 686 | 99 | |
NM_019283.3 | NP_062156.2 | 3 - 4 | 811 | 99 | ||
GenBank mRNA | AB015433.1 | - | 3 - 4 | 777 | 99 | |
BC061989.1 | - | 2 - 3 | 669 | 99 | ||
FQ148223.1 | - | 2 - 3 | 686 | 99 | ||
U59324.1 | - | 2 - 3 | 600 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
